Anti-IL17RB Rat Monoclonal Antibody [clone: 752101]

Supplier: R & D Systems
MAB10402
102885-224EA 577.02 USD
102885-224
Anti-IL17RB Rat Monoclonal Antibody [clone: 752101]
Antibodies
Type: Primary
Antigen: IL17RB
Clonality: Monoclonal
Clone: 752101
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR