Anti-IL17RB Rat Monoclonal Antibody [clone: 152309]

Supplier: R & D Systems
MAB10401
220018-733EA 772.25 USD
220018-733
Anti-IL17RB Rat Monoclonal Antibody [clone: 152309]
Antibodies
Type: Primary
Antigen: IL17RB
Clonality: Monoclonal
Clone: 152309
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR