Anti-CD244 Mouse Monoclonal Antibody [clone: 146510]

Supplier: R & D Systems
MAB1039
220018-731EA 772.25 USD
220018-731
Anti-CD244 Mouse Monoclonal Antibody [clone: 146510]
Antibodies
Type: Primary
Antigen: CD244
Clonality: Monoclonal
Clone: 146510
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR