Anti-STRO1 Mouse Monoclonal Antibody [clone: STRO-1]

Supplier: R & D Systems
MAB1038
220018-730EA 647.88 USD
220018-730
Anti-STRO1 Mouse Monoclonal Antibody [clone: STRO-1]
Antibodies
Type: Primary
Antigen: STRO1
Clonality: Monoclonal
Clone: STRO-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR