Anti-IL19 Mouse Monoclonal Antibody [clone: 152107]

Supplier: R & D Systems
MAB10351
220018-728EA 772.25 USD
220018-728
Anti-IL19 Mouse Monoclonal Antibody [clone: 152107]
Antibodies
Type: Primary
Antigen: IL19
Clonality: Monoclonal
Clone: 152107
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR