Anti-ADAM8 Mouse Monoclonal Antibody [clone: 143338]

Supplier: R & D Systems
MAB10311
220018-725EA 763.57 USD
220018-725
Anti-ADAM8 Mouse Monoclonal Antibody [clone: 143338]
Antibodies
Type: Primary
Antigen: ADAM8
Clonality: Monoclonal
Clone: 143338
Conjugation: Unconjugated
Epitope: 498-653 aa
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR