Anti-CTSD Rat Monoclonal Antibody [clone: 204712]

Supplier: R & D Systems
MAB1029
220018-723EA 779.48 USD
220018-723
Anti-CTSD Rat Monoclonal Antibody [clone: 204712]
Antibodies
Type: Primary
Antigen: CTSD
Clonality: Monoclonal
Clone: 204712
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR