Anti-IFNGR1 Rat Monoclonal Antibody [clone: 170911]

Supplier: R & D Systems
MAB10262
220018-720EA 772.25 USD
220018-720
Anti-IFNGR1 Rat Monoclonal Antibody [clone: 170911]
Antibodies
Type: Primary
Antigen: IFNGR1
Clonality: Monoclonal
Clone: 170911
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR