Anti-BTC Rat Monoclonal Antibody [clone: 197415]

Supplier: R & D Systems
MAB1025
220018-716EA 763.57 USD
220018-716
Anti-BTC Rat Monoclonal Antibody [clone: 197415]
Antibodies
Type: Primary
Antigen: BTC
Clonality: Monoclonal
Clone: 197415
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR