Anti-CCL4 Mouse Monoclonal Antibody [clone: 163910]

Supplier: R & D Systems
MAB1024
220018-715EA 763.57 USD
220018-715
Anti-CCL4 Mouse Monoclonal Antibody [clone: 163910]
Antibodies
Type: Primary
Antigen: CCL4
Clonality: Monoclonal
Clone: 163910
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Cotton Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR