Anti-PDCD1LG2 Rat Monoclonal Antibody [clone: 168633]

Supplier: R & D Systems
MAB1022
220018-714EA 594.37 USD
220018-714
Anti-PDCD1LG2 Rat Monoclonal Antibody [clone: 168633]
Antibodies
Type: Primary
Antigen: PDCD1LG2
Clonality: Monoclonal
Clone: 168633
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR