Anti-IL2RA Mouse Monoclonal Antibody [clone: 24212]

Supplier: R & D Systems
MAB1020
220018-713EA 779.48 USD
220018-713
Anti-IL2RA Mouse Monoclonal Antibody [clone: 24212]
Antibodies
Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: 24212
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR