Anti-CD274 Rat Monoclonal Antibody [clone: 179711]

Supplier: R & D Systems
MAB1019
220018-712EA 633.42 USD
220018-712
Anti-CD274 Rat Monoclonal Antibody [clone: 179711]
Antibodies
Type: Primary
Antigen: CD274
Clonality: Monoclonal
Clone: 179711
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR