Anti-CTSD Mouse Monoclonal Antibody [clone: 185111]

Supplier: R & D Systems
MAB1014
220018-710EA 763.57 USD
220018-710
Anti-CTSD Mouse Monoclonal Antibody [clone: 185111]
Antibodies
Type: Primary
Antigen: CTSD
Clonality: Monoclonal
Clone: 185111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR