Anti-CTSH Rat Monoclonal Antibody [clone: 175909]

Supplier: R & D Systems
MAB1013
220018-709EA 763.57 USD
220018-709
Anti-CTSH Rat Monoclonal Antibody [clone: 175909]
Antibodies
Type: Primary
Antigen: CTSH
Clonality: Monoclonal
Clone: 175909
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR