Anti-TNFA Mouse Monoclonal Antibody [clone: 159813]

Supplier: R & D Systems
MAB10111
220018-708EA 763.57 USD
220018-708
Anti-TNFA Mouse Monoclonal Antibody [clone: 159813]
Antibodies
Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: 159813
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cotton Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR