Anti-TNFA Mouse Monoclonal Antibody [clone: 159811]

Supplier: R & D Systems
MAB1011
220018-707EA 763.57 USD
220018-707
Anti-TNFA Mouse Monoclonal Antibody [clone: 159811]
Antibodies
Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: 159811
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Cotton Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR