Anti-LTBR Rat Monoclonal Antibody [clone: 157105]

Supplier: R & D Systems
MAB1008
220018-705EA 763.57 USD
220018-705
Anti-LTBR Rat Monoclonal Antibody [clone: 157105]
Antibodies
Type: Primary
Antigen: LTBR
Clonality: Monoclonal
Clone: 157105
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR