Anti-UNC5B Mouse Monoclonal Antibody [clone: 187305]

Supplier: R & D Systems
MAB1006
220018-703EA 763.57 USD
220018-703
Anti-UNC5B Mouse Monoclonal Antibody [clone: 187305]
Antibodies
Type: Primary
Antigen: UNC5B
Clonality: Monoclonal
Clone: 187305
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR