Anti-UNC5C Mouse Monoclonal Antibody [clone: 200414]

Supplier: R & D Systems
MAB1005
220018-702EA 763.57 USD
220018-702
Anti-UNC5C Mouse Monoclonal Antibody [clone: 200414]
Antibodies
Type: Primary
Antigen: UNC5C
Clonality: Monoclonal
Clone: 200414
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR