Anti-IL1RL1 Rat Monoclonal Antibody [clone: 245714]

Supplier: R & D Systems
MAB1004
220018-700EA 763.57 USD
220018-700
Anti-IL1RL1 Rat Monoclonal Antibody [clone: 245714]
Antibodies
Type: Primary
Antigen: IL1RL1
Clonality: Monoclonal
Clone: 245714
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR