Anti-HA (Peptide) Mouse Monoclonal Antibody [clone: 543851]

Supplier: R & D Systems
MAB060
102885-222EA 655.11 USD
102885-222
Anti-HA (Peptide) Mouse Monoclonal Antibody [clone: 543851]
Antibodies
Type: Primary
Antigen Symbol: HA (Peptide)
Antigen Name: Hemagglutinin
Antigen Synonyms:
Clonality: Monoclonal
Clone: 543851
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR