Anti-6xHis Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: AD1.1.1]

Supplier: R & D Systems
MAB050H
220018-692EA 639.2 USD
220018-692
Anti-6xHis Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: AD1.1.1]
Antibodies
Simple Western

Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: AD1.1.1
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR