Anti-Hamster IgG Mouse Monoclonal Antibody [clone: MAH1.12]

Supplier: R & D Systems
MAB011
220018-690EA 326.83 USD
220018-690
Anti-Hamster IgG Mouse Monoclonal Antibody [clone: MAH1.12]
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: MAH1.12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR