Anti-TIMP3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 277128]

Supplier: R & D Systems
IC9731A
220018-509EA 594.37 USD
220018-509
Anti-TIMP3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 277128]
Antibodies
Type: Primary
Antigen: TIMP3
Clonality: Monoclonal
Clone: 277128
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR