Anti-SPARC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 122511]

Supplier: R & D Systems
IC941P
220018-506EA 513.38 USD
220018-506
Anti-SPARC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 122511]
Antibodies
Type: Primary
Antigen: SPARC
Clonality: Monoclonal
Clone: 122511
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR