Anti-MMP9 Mouse Monoclonal Antibody (Fluorescein) [clone: 56129]

Supplier: R & D Systems
IC9111F
220018-499EA 454.09 USD
220018-499
Anti-MMP9 Mouse Monoclonal Antibody (Fluorescein) [clone: 56129]
Antibodies
Type: Primary
Antigen: MMP9
Clonality: Monoclonal
Clone: 56129
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR