Anti-MMP2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1A10]

Supplier: R & D Systems
IC9023P
220018-496EA 549.54 USD
220018-496
Anti-MMP2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1A10]
Antibodies
Type: Primary
Antigen: MMP2
Clonality: Monoclonal
Clone: 1A10
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR