Anti-TCF7 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 812145]

Supplier: R & D Systems
IC8224G
102884-814EA 582.8 USD
102884-814
Anti-TCF7 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 812145]
Antibodies
Type: Primary
Antigen: TCF7
Clonality: Monoclonal
Clone: 812145
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR