Anti-FOXP3 Rabbit Monoclonal Antibody (APC (Allophycocyanin)) [clone: 1054C]

Supplier: R & D Systems
IC8214A-100
102884-806EA 480.12 USD
102884-806
Anti-FOXP3 Rabbit Monoclonal Antibody (APC (Allophycocyanin)) [clone: 1054C]
Antibodies
Type: Primary
Antigen Symbol: FOXP3
Antigen Name: forkhead box P3
Antigen Synonyms: JM2, scurfin, sf
Clonality: Monoclonal
Clone: 1054C
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR