Anti-VIM Goat Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
IC8104G
102884-804EA 561.11 USD
102884-804
Anti-VIM Goat Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: VIM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR