Anti-BATF Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 687706]

Supplier: R & D Systems
IC8054A
102884-802EA 594.37 USD
102884-802
Anti-BATF Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 687706]
Antibodies
Type: Primary
Antigen: BATF
Clonality: Monoclonal
Clone: 687706
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR