Anti-EEA1 Sheep Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
IC8047G
102884-800EA 594.37 USD
102884-800
Anti-EEA1 Sheep Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: EEA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR