Anti-ISG15 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 851701]

Supplier: R & D Systems
IC8044P
102884-798EA 566.89 USD
102884-798
Anti-ISG15 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 851701]
Antibodies
Type: Primary
Antigen: ISG15
Clonality: Monoclonal
Clone: 851701
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR