Anti-SQSTM1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 864807]

Supplier: R & D Systems
IC8028G
102884-792EA 577.02 USD
102884-792
Anti-SQSTM1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 864807]
Antibodies
Type: Primary
Antigen: SQSTM1
Clonality: Monoclonal
Clone: 864807
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR