Anti-ARG1 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 658922]

Supplier: R & D Systems
IC8026N
102884-788EA 594.37 USD
102884-788
Anti-ARG1 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 658922]
Antibodies
Type: Primary
Antigen: ARG1
Clonality: Monoclonal
Clone: 658922
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR