Anti-CXCL13 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 53610]

Supplier: R & D Systems
IC801A
220018-490EA 600.15 USD
220018-490
Anti-CXCL13 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 53610]
Antibodies
Type: Primary
Antigen: CXCL13
Clonality: Monoclonal
Clone: 53610
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR