Anti-PF4 Mouse Monoclonal Antibody (Fluorescein) [clone: 170138]

Supplier: R & D Systems
IC7952F
220018-488EA 438.18 USD
220018-488
Anti-PF4 Mouse Monoclonal Antibody (Fluorescein) [clone: 170138]
Antibodies
Type: Primary
Antigen: PF4
Clonality: Monoclonal
Clone: 170138
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR