Anti-ERK1 + ERK2 Rabbit Monoclonal Antibody (Alexa Fluor® 488) [clone: 269434]

Supplier: R & D Systems
IC7806G
102884-748EA 577.02 USD
102884-748
Anti-ERK1 + ERK2 Rabbit Monoclonal Antibody (Alexa Fluor® 488) [clone: 269434]
Antibodies
Type: Primary
Antigen: ERK1 + ERK2
Clonality: Monoclonal
Clone: 269434
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR