Anti-AKT Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5457]

Supplier: R & D Systems
IC7794P
102884-746EA 566.89 USD
102884-746
Anti-AKT Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5457]
Antibodies
Type: Primary
Antigen: AKT
Clonality: Monoclonal
Clone: 5457
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR