Anti-IL1F10 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 798036]

Supplier: R & D Systems
IC7774G
102884-742EA 519.17 USD
102884-742
Anti-IL1F10 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 798036]
Antibodies
Type: Primary
Antigen: IL1F10
Clonality: Monoclonal
Clone: 798036
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR