Anti-CK18 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 810811]

Supplier: R & D Systems
IC7619G
102884-736EA 561.11 USD
102884-736
Anti-CK18 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 810811]
Antibodies
Type: Primary
Antigen: CK18
Clonality: Monoclonal
Clone: 810811
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR