Anti-NLRP3 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 768319]

Supplier: R & D Systems
IC7578G
102884-724EA 594.37 USD
102884-724
Anti-NLRP3 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 768319]
Antibodies
Type: Primary
Antigen: NLRP3
Clonality: Monoclonal
Clone: 768319
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR