Anti-BATF3 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 841702]

Supplier: R & D Systems
IC7437G
102884-302EA 561.11 USD
102884-302
Anti-BATF3 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 841702]
Antibodies
Type: Primary
Antigen: BATF3
Clonality: Monoclonal
Clone: 841702
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR