Anti-SNAI2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 666633]

Supplier: R & D Systems
IC7408P
102884-296EA 543.75 USD
102884-296
Anti-SNAI2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 666633]
Antibodies
Type: Primary
Antigen: SNAI2
Clonality: Monoclonal
Clone: 666633
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR