Anti-BrdU Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: BU-1]

Supplier: R & D Systems
IC7225A
102884-290EA 633.42 USD
102884-290
Anti-BrdU Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: BU-1]
Antibodies
Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone: BU-1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR