Anti-RET Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 132507]

Supplier: R & D Systems
IC718A
220018-480EA 577.02 USD
220018-480
Anti-RET Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 132507]
Antibodies
Type: Primary
Antigen: RET
Clonality: Monoclonal
Clone: 132507
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR