Anti-ESR2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 733930]

Supplier: R & D Systems
IC7106G
102884-282EA 561.11 USD
102884-282
Anti-ESR2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 733930]
Antibodies
Type: Primary
Antigen: ESR2
Clonality: Monoclonal
Clone: 733930
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR