Anti-AXL Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 713610]

Supplier: R & D Systems
IC6965G
102884-488EA 552.43 USD
102884-488
Anti-AXL Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 713610]
Antibodies
Type: Primary
Antigen: AXL
Clonality: Monoclonal
Clone: 713610
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR