Anti-IL2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 100312]

Supplier: R & D Systems
IC6521P
220018-476EA 535.08 USD
220018-476
Anti-IL2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 100312]
Antibodies
Type: Primary
Antigen: IL2
Clonality: Monoclonal
Clone: 100312
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR